((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-((methylsulfonyl)oxy)tetrahydrofuran-2-yl)methyl benzoate 98%

Supplier: Ambeed
Warning

A152709-250MG A152709-1G A152709-25G A152709-5G
103804-422EA 20 USD
103804-422 103842-594 77110-382 77299-968 77299-970 77299-972
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-((methylsulfonyl)oxy)tetrahydrofuran-2-yl)methyl benzoate 98%
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-((methylsulfonyl)oxy)tetrahydrofuran-2-yl)methyl benzoate

Precaución: for Research Use Only

Formula: C₂₀H₁₈F₂O₈S
MW: 456.41 g/mol
Boiling Pt: 588.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08460089
CAS Number: 122111-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR