Benzyl-2-bromoethyl ether 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
((2-Bromoethoxy)methyl)benzene, Benzyloxyethyl Bromide, (2-Bromoethoxymethyl)benzene
A249841-1000G
A249841-100G
A249841-10G
A249841-500G
A249841-25G
A249841-5G
A249841-1G
103811-894EA
5
USD
103811-894
103851-732
76692-454
76692-456
76692-458
76692-460
77091-080
77091-082
77304-502
Benzyl-2-bromoethyl ether 97%
Benzyl-2-bromoethyl ether
Precaución: for Research Use Only
Formula:
C₉H₁₁BrO MW: 215.09 g/mol Boiling Pt: 258.6 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD01321307 CAS Number: 1462-37-9 UN: 3334 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...