((1S,4R)-4-Aminocyclopent-2-en-1-yl)methanol hydrochloride 95%

Supplier: Ambeed
Warning

A283589-1G A283589-25G A283589-5G A283589-100G A283589-10G A283589-250MG A283589-500G
103813-618EA 6 USD
103813-618 103853-568 76837-276 76837-278 76837-280 77166-656 77166-658 77166-660 77683-853
((1S,4R)-4-Aminocyclopent-2-en-1-yl)methanol hydrochloride 95%
((1S,4R)-4-Aminocyclopent-2-en-1-yl)methanol hydrochloride

Precaución: For research use only.

Formula: C₆H₁₂ClNO
MW: 149.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632106
CAS Number: 168960-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR