((1S,4R)-4-(2-Amino-6-chloro-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hydrochloride 95%

Supplier: Ambeed
Danger

A508327-1G A508327-25G A508327-5G
103828-354EA 6 USD
103828-354 76843-436 77017-284 77017-286 77891-032 77891-033
((1S,4R)-4-(2-Amino-6-chloro-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hydrochloride 95%
((1S,4R)-4-(2-Amino-6-chloro-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃Cl₂N₅O
Boiling Pt: 572 °C (760 mm Hg)
Flash Pt: 299.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12546177
CAS Number: 172015-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR