((1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetrakis(methylene))tetraphosphonic acid 98%

Supplier: Ambeed
Warning

Synonyms: DOTMP

A100369-100MG A100369-1G A100369-250MG
103795-394EA 91 USD
103795-394 76821-200 77296-650 77682-929
((1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetrakis(methylene))tetraphosphonic acid 98%
((1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetrakis(methylene))tetraphosphonic acid

Precaución: For research use only.

Formula: C₁₂H₃₂N₄O₁₂P₄
MW: 548.29 g/mol
Boiling Pt: 921.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01863112
CAS Number: 91987-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR