Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
P7717L
P7717S
76326-040EA
594
USD
76326-040
76326-042
Unstained Protein Standard, Broad Range, New England Biolabs
Standards
Organic Standards
The unstained protein standard, broad range is a mixture of highly pure, recombinant proteins that resolves into 13 sharp bands from 10 to 200 kDa when electrophoresed and stained with Coomassie Brilliant Blue R-250 or other commercially available, ready-to-use protein stains.
- Allows accurate molecular weight determination when performing SDS-PAGE analysis
- Direct loading, additional loading buffer and heat incubation not required
- Recombinant proteins with no detectable protease contaminating activities
- Optimal stability for up to 24 months