GelGreen® Agarose LE, Biotium
Supplier: Biotium
41030-50G 41030-5G
76414-952EA 317.39 USD
76414-952 76414-954
GelGreen® Agarose LE, Biotium
Electrophoresis Gels Agarose and Acrylamide
Ultrapure molecular biology grade agarose pre-coated with GelGreen® Nucleic acid gel stain for convenient and safer preparation of precast gels.
  • Non-toxic GelGreen® DNA dye is already in the agarose
  • No need to handle concentrated dye solutions, for enhanced safety
  • Use with TAE or TBE for 0.8 to 2% gels
  • Low EEO, ultrapure molecular biology grade agarose
  • Compatible with blue light illuminators
GelGreen® Agarose LE is our ultra-pure molecular biology grade LE agarose pre-coated with GelGreen® Nucleic acid gel stain. There is no need to handle concentrated fluorescent dye while preparing your gel, for greater convenience and safety. For agarose gels from 0.8 to 2% in TAE or TBE.
GelGreen® is a sensitive, stable and environmentally safe green fluorescent nucleic acid dye specifically designed for gel staining. GelGreen® is far more sensitive than SYBR® Safe. Unlike SYBR® dyes, which are known to be unstable, GelGreen® is very stable, both hydrolytically and thermally. GelGreen® has UV absorption between 250 and 300 nm and a strong absorption peak centered around 500 nm.