MicroMax® VP Disposable Protective Coveralls, Lakeland Industries
Supplier: Lakeland
MVP414-LG MVP414-XL MVP414-2X MVP414-3X MVP414-4X MVP428-LG MVP428-XL MVP428-2X MVP428-3X MVP428-4X
76449-784CS 371.71 USD
76449-784 76449-786 76449-788 76449-790 76449-792 76449-794 76449-796 76449-798 76449-800 76449-802
MicroMax® VP Disposable Protective Coveralls, Lakeland Industries
Disposable Garments
MicroMax®VP garments are specifically designed to protect when the risk of blood, body fluids, bloodborne pathogens, and viral contamination are the greatest. Ideal for use in crime labs, crime scene cleanup, and by emergency response personnel.
  • Protective hood
  • Seamless front reduces risk of contaminant exposure
  • Taped storm flap protects zipper
  • Elastic back for more comfortable fit
  • Material: 55 g SBPP with laminated microporous film
  • Passes ASTM F1670/F1671 for Blood and Viral Protection
From the fabric composition, to the garment design, the MicroMax® VP family of products are ideal for use when workers may come into contact with bloodborne pathogens. The seamless front reduces the risk of possible contamination from liquid penetration, while a taped storm flap keeps contaminants away from the zipper. An elastic back ensures a more comfortable fit and helps prevent rip-outs. These coveralls feature a protective hood, and are also available with attached boots to further prevent cross-contamination.
Certificaciones: ASTM F1670. ASTM F1671.