Mouse Recombinant Siglec-10 (from HEK293 cells)

Supplier: ACROBIOSYSTEMS INC MS

SI0M52H71MG SI0M52H7100UG
103815-060EA 2929.23 USD
103815-060 103815-062
Mouse Recombinant Siglec-10 (from HEK293 cells)
Proteins and Peptides

Mouse Siglec-10 Protein, His Tag, ACROBiosystems

Order Now

SPECIFICATIONS

Protein/Peptide Name Siglec-10
Protein Synonyms Siglec10|PRO940|SIGLEC10|SLG2|MGC126774
Environmentally Preferable
Protein/Peptide Type Recombinant
Species Mouse
Source HEK293 cells
Conjugation Unconjugated
Gene ID Q80ZE3-1
Endotoxin Level Low
Endotoxin Content Less than 1.0 EU per μg by the LAL method.
Purity >90% as determined by SDS-PAGE.
Formulation Lyophilized from 0.22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH5.5. Normally trehalose is added as protectant before lyophilization.
Molecular Weight 60.7 kDa
Storage Conditions –20 °C
Shipping Temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR