Cynomolgus monkey Recombinant CD27 Ligand (from HEK293 cells)

Supplier: ACROBIOSYSTEMS INC MS

CDLC52441MG CDLC5244100UG
103871-042EA 2487.69 USD
103871-042 103871-044
Cynomolgus monkey Recombinant CD27 Ligand (from HEK293 cells)
Proteins and Peptides

Cynomolgus CD27 Ligand / CD70 Protein, His Tag (active trimer) (MALS verified), ACROBiosystems

Order Now

SPECIFICATIONS

Protein/Peptide Name CD27 Ligand
Protein Synonyms CD27LG|TNFSF7|TNFSF7G|CD27L|CD70
Environmentally Preferable
Protein/Peptide Type Recombinant
Species Cynomolgus monkey
Source HEK293 cells
Conjugation Unconjugated
Gene ID G7PYU6-1
Endotoxin Level Low
Endotoxin Content Less than 1.0 EU per μg by the LAL method.
Purity >90% as determined by SDS-PAGE.
Formulation Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally Trehalose is added as protectant before lyophilization.
Molecular Weight 19.2 kDa
Storage Conditions –20 °C
Shipping Temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR