2-(Tetrazol-5-yl)phenylboronic acid 97%

Supplier: Ambeed
Danger

Synonyms: 2-(5-Tetrazolyl)phenylboronic acid, (2-(1H-Tetrazol-5-yl)phenyl)boronic acid, Boronic acid,B-[2-(2H-tetrazol-5-yl)phenyl]-, 2-(1H-Tetrazol-5-yl)phenylboronic acid

A100009-250MG A100009-1G A100009-5G
103795-194EA 45 USD
103795-194 103820-916 77008-254 77296-530 77296-532
2-(Tetrazol-5-yl)phenylboronic acid 97%
2-(Tetrazol-5-yl)phenylboronic acid

  • Building blocks & intermediates
  • High-quality
  • Competitive price
  • Fast delivery


Precaución: for Research Use Only

Formula: C₇H₇BN₄O₂
MW: 189.97 g/mol
Boiling Pt: 507.1 °C (760 mm Hg)
Flash Pt: 260.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06796325
CAS Number: 155884-01-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR