4-(Trifluoromethyl)benzylamine 98%
Supplier: Ambeed
|
Warning
|
A100008-10G
A100008-25G
A100008-1G
A100008-5G
A100008-100G
103795-192EA
5
USD
103795-192
103820-915
76686-962
76686-964
77296-526
77296-528
77623-800
4-(Trifluoromethyl)benzylamine 98%
4-(Trifluoromethyl)benzylamine
- Building Blocks & Intermediates
- High-quality
- Competitive Price
- Fast Delivery
Precaución: for Research Use Only
Formula:
C₈H₈F₃N Boiling Pt: 191 °C (760 mm Hg) Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010220 CAS Number: 3300-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...