Anti-FMC63 Hybridoma Polyclonal Antibody(FITC Labeled Protein)

Supplier: ACROBIOSYSTEMS INC MS
FM3FY45200TESTS FM3FY4525TESTS
103871-452EA 1976.15 USD
103871-452 103871-454
Anti-FMC63 Hybridoma Polyclonal Antibody(FITC Labeled Protein)
Antibodies
Type: Primary
Antigen: FMC63
Clonality: Polyclonal
Clone:
Conjugation: FITC Labeled Protein
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR