Anti-Nucleocapsid protein HEK293 (Mouse) Polyclonal Antibody (AS47) (Powder)

Supplier: ACROBIOSYSTEMS INC MS
NUNS47A11MG NUNS47A1100UG
103871-006EA 2692.31 USD
103871-006 103871-008
Anti-Nucleocapsid protein HEK293 (Mouse) Polyclonal Antibody (AS47) (Powder)
Antibodies
Type: Primary
Antigen: Nucleocapsid protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Mouse IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR