Anti-Nucleocapsid Protein Hybridoma Polyclonal Antibody (Trehalose free) (AS57)

Supplier: ACROBIOSYSTEMS INC MS
NUNS571MG NUNS57100UG
103870-936EA 2692.31 USD
103870-936 103870-938
Anti-Nucleocapsid Protein Hybridoma Polyclonal Antibody (Trehalose free) (AS57)
Antibodies
Type: Primary
Antigen: Nucleocapsid protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: Mouse IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR