Poly(3-hexylthiophene-2,5-diyl), Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
P3HT, Poly(3-hexylthiophene), Poly[3-hexylthiophene-2,5-diyl]
H66726-03
H66726-ME
AAH66726-03EA
715.45
USD
AAH66726-03
AAH66726-ME
Poly(3-hexylthiophene-2,5-diyl), Organic Photovoltaics (OPV) reagent
Poly(3-hexylthiophene-2,5-diyl)
MW:
196.352 g/mol |
MDL Number:
MFCD00217686 CAS Number: 104934-50-1 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | T.b.d. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...