BactoView™ Live Fluorescent Bacterial Stains, Biotium
Supplier: Biotium
40101 40101-T 40102 40102-T
76403-972EA 351.07 USD
76403-972 76403-974 76403-976 76403-978
BactoView™ Live Fluorescent Bacterial Stains, Biotium
Stains Microbiology Stains
Live cell bacteria stains suitable for flow cytometry or fluorescence microscopy. BactoView™ Live dyes can stain live and dead, gram-positive and gram-negative bacteria with green or red fluorescence.
  • DNA dyes for bacteria
  • Stain live and dead, gram-positive and gram negative cells
  • Choose green or red fluorescence
  • Supplied at 500X in DMSO
BactoView™ Live dyes are fluorogenic live cell bacterial DNA stains. Bright and cell-permeant, they are able to stain both live and dead gram-positive and gram-negative bacteria for microscopy or flow cytometry.
BactoView™ Live dyes can stain both live and dead bacteria; dead cells tend to stain more brightly than live cells. BactoView™ Live Green can stain mammalian cell nuclei, but BactoView™ Live Red shows weak mitochondrial signal in mammalian cells. In yeast, staining with either BactoView™ Live dye is weak and not nuclear.