Antimicrobial Charging Carts, Tripp Lite
Supplier: Tripp Lite
CSC32ACWHG CSC32USBWHG
76461-430EA 4366.43 USD
76461-430 76461-432
Antimicrobial Charging Carts, Tripp Lite
Carts
Charges, secures, organizes and sanitizes chromebooks and laptops or iPad and android tablets.
  • Mobile charging cart accommodates numerous electronics
  • UV light helps sanitize devices without potentially dangerous heat, liquids or chemicals
  • Exclusive, patented antimicrobial protection fights surface bacteria
  • Locking steel enclosure keeps valuable equipment safe and secure
  • Moving your devices from room to room is easy
  • Well-organized to avoid confusing cable clutter
The charging stations provide AC or USB charging, secure storage, cord management and surface sanitization for as many as 32 devices. A built-in circuit breaker protects the devices against overloads. The charging station is recommended for healthcare facilities, classrooms, businesses, government agencies, restaurants, hotels and anywhere else that uses mobile devices on a daily basis.
The charging cart helps reduce the presence of germs on Chromebooks, laptops and tablets. Built-in ultraviolet lights help disinfect electronic devices at the touch of a button by destroying most microbes without the use of damaging heat, liquids or chemicals. The germicidal UVC bulbs are on a timer to control the amount of UV exposure the devices receive. An LED on the exterior of the cabinet indicates when the doors are closed and the start button can be pressed to activate the 10-minute timer. A second LED indicates when the UVC lights are on. If a door is opened during sanitization, the lights automatically shut off for safety.
The charging cart's interior and exterior surfaces, including the handle, are protected by a patented antimicrobial powder coating with EPA-registered and FDA-compliant silver ionic technology. The antimicrobial surface protection is 99.9 percent effective in inhibiting the growth of bacteria and viruses, such as C. diff and MRSA. The coating meets the requirements of JIS Z 2801:2000, an international standard for evaluating efficacy in antimicrobial products.
Four pre-installed swivel casters with large non-marking wheels improve the ride over uneven surfaces, including carpeting and thresholds, and lock to help prevent unwanted movement. The reversible wide-grip handle makes the cart easy to control.
Cables and connectors run through strain relief bushings, keeping them organized and out of the way, and are protected by blanking panels. Adjustable dividers accommodate up to 16 devices per shelf. Front doors open at least 180° to allow maximum access to the interior. A power cord wrap keeps the input cord out of the way when moving the charging station. Access grommets allow the 10 ft. (3.05 m) cord with hospital-grade NEMA 5-15P plug to extend through either side of the cart, allowing great flexibility in reaching outlets.
Certificaciones: Tested to EIA-310-E, UL60950, FCC & IEC Class B.