Potassium citrate monohydrate 99.0-100.5% (dry basis), meets analytical specification of USP, Sigma-Aldrich®

Synonyms: Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate

P1722-100G P1722-500G
220201-494EA 43.38 USD
220201-494 220201-495
Potassium citrate monohydrate 99.0-100.5% (dry basis), meets analytical specification of USP, Sigma-Aldrich®
Potassium citrate monohydrate
Formula: HOC(COOK)(CH₂COOK)₂·H₂O
MW: 324.41 g/mol
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5

Order Now

Specification Test Results

Loss on Drying 3.0 - 6.0%
Identity Pass
Alkalinity Pass
Assay (Dry Basis) 99.0 - 100.5 %
Tartrate Pass
Residual Solvents USP Meets Requirements

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR