Huma OsteoGelatin High Bloom Gelatin, Lyophilized, Humabiologics
Supplier: HUMABIOLOGICS, INC
BGHL-100MG BGHL-250MG BGHL-500MG BGHL-1G BGHL-5G
76452-642EA 74.13 USD
76452-642 76452-644 76452-646 76452-648 76452-650
Huma OsteoGelatin High Bloom Gelatin, Lyophilized, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human bone gelatin for hydrogel and bioink applications.
  • Native human bone gelatin
  • Clinically relevant and xeno-free for regenerative medicine applications
  • Lyophilized for tailored 3D hydrogel or bioink applications
  • Supports various cell attachments
  • Bloom strength is 225 to 325 g
Huma OsteoGelatin is isolated from human bone sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. Huma OsteoGelatin has undergone a proprietary manufacturing method to extract human gelatin from bone. Huma OsteoGelatin bone-derived gelatin is supplied lyophilized for hydrogel, bioink, or cell culture applications.
Huma OsteoGelatin addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of Huma OsteoGelatin has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
Información para pedidos: Huma OsteoGelatin High Bloom is available in 100, 250, 500 mg and 1 and 5 g (5×1 g).
Precaución: Huma OsteoGelatin is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.