DL-Glyceraldehyde dimer ≥95%

Synonyms: 3,6-Dihydroxy-1,4-dioxane-2,5-dimethanol, (2R,3S,5S,6R)-3,6-Bis(hydroxymethyl)-1,4-dioxane-2,5-diol

G4802-5G G4802-1G
220179-336EA 1098.46 USD
220179-336 220179-335
DL-Glyceraldehyde dimer ≥95%
DL-Glyceraldehyde dimer
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00070528
CAS Number: 23147-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR