Isopropylmagnesium chloride - LiCl complex solution 1.3 M in THF, Sigma-Aldrich®

Danger

Synonyms: Isopropylmagnesium chloride - Lithium chloride complex, Turbo Grignard

656984-800ML 656984-100ML 656984-4X25ML
220085-778EA 258.23 USD
220085-778 220085-777 102299-978
Isopropylmagnesium chloride - LiCl complex solution 1.3 M in THF, Sigma-Aldrich®
Isopropylmagnesium chloride - LiCl complex
Formula: C₃H₇ClMg·LiCl
MW: 145.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784514
CAS Number: 807329-97-1
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR