Methyl viologen dichloride hydrate ≥98%, Sigma-Aldrich®

Danger

Synonyms: Methyl Viologen hydrate, Gramoxone, Paraquat dichloride hydrate, 1,1′-Dimethyl-4,4′-bipyridinium dichloride

856177-1G 856177-250MG
220098-308EA 80.03 USD
220098-308 220098-309
Methyl viologen dichloride hydrate ≥98%, Sigma-Aldrich®
Methyl viologen dichloride hydrate
Formula: C₁₂H₁₄Cl₂N₂·xH₂O
MW: 257.16 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150001
CAS Number: 75365-73-0
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR