4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride 97%

Synonyms: 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride, 4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride, (4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine hydrochloride, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine hydrochloride, 4-Aminomethylphenylboronic acid pinacol ester HCl

M-Clarity™ quality level = MQ100

741213-500MG 741213-1G
102299-990EA 68.4 USD
102299-990 102300-082
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride 97%
4-(Aminomethyl)phenylboronic acid pinacol ester hydrochloride
Formula: C₁₃H₂₀BNO₂·HCl
MW: 269.58 g/mol
Melting Pt: 254.1…262.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179455
CAS Number: 850568-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR