Westex® DH 6.5 oz. Men's FR Button Up Shirts, Hi-Vis Yellow, Lakeland Industries
Supplier: Lakeland
ISH65DH29RT-2X ISH65DH29RT-2XT ISH65DH29RT-3X ISH65DH29RT-3XT ISH65DH29RT-4X ISH65DH29RT-4XT ISH65DH29RT-5X ISH65DH29RT-5XT ISH65DH29RT-6X ISH65DH29RT-6XT ISH65DH29RT-LG ISH65DH29RT-LGT ISH65DH29RT-MD ISH65DH29RT-MDT ISH65DH29RT-SM ISH65DH29RT-XL ISH65DH29RT-XLT ISH65DH29RT-XS
76438-678EA 204.64 USD
76438-678 76438-680 76438-682 76438-684 76438-686 76438-688 76438-690 76438-692 76438-694 76438-696 76438-698 76438-700 76438-702 76438-704 76438-706 76438-708 76438-710 76438-712
Westex® DH 6.5 oz. Men's FR Button Up Shirts, Hi-Vis Yellow, Lakeland Industries
Workwear
The 6.5 oz. Westex® DH Men's Button Up Shirt has a uniquely woven fabric blend that provides outstanding breathability and advanced moisture management to keep the wearer cooler, drier, and more comfortable.
  • Permanent moisture wicking properties
  • Lightweight 6.5 oz. Westex® DH inherently FR fabric
  • Tag-free collar
  • Athletic cut
  • Button down collar
  • Drop shoulder design
  • Long rounded tail stays tucked in
Mid layers are the versatile garments that can help take the chill off for a few hours on a fall morning, or provide an extra layer of insulation for those long, frigid winter days. Paired with a Base layer, Mid layers continue to pull moisture away from the body and move it to the outermost garment so it can dry. Lightweight, effective fabrics let Mid layers increase comfort without reducing mobility.
Certificaciones: NFPA 70E. NFPA 2112 - UL certified. APTV: 8.9 cal/cm². CAT: 2. ANSI 107 Class 3.