Di-n-hexyl phthalate Single-Component Organic Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
S-4155-H CRM-PE-DNHP S-4155
SXS-4155-HEA 101.56 USD
SXS-4155-H SXCRM-PE-DNHP SXS-4155
Di-n-hexyl phthalate Single-Component Organic Standard, SPEX CertiPrep
Standards Organic Standards
Consumer safety standards, phthalates in polyethylene QC standards.
Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.