Calcium (Ca2+) Single-Element Ion Cation Standard, SPEX CertiPrep
Supplier: SPEX CertiPrep
CS-CA2-2Y
SXCS-CA2-2YEA 87.2 USD
SXCS-CA2-2Y
Calcium (Ca2+) Single-Element Ion Cation Standard, SPEX CertiPrep
Standards Cationic Standards Calcium Standards
Single-element standards for ion chromatography and ion selective electodes.
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.