Calcium chloride dihydrate USP
Supplier: RPI
|
Warning
|
Synonyms:
Calcium Chloride Dihydrate, Calcium dichloride
C36200-1000.0
C36200-500.0
C36200-5000.0
76345-432EA
96.64
USD
76345-432
76345-434
76345-436
Calcium chloride dihydrate USP
Calcium chloride dihydrate
Formula:
CaCl₂·2H₂O MW: 147.01 g/mol Boiling Pt: 1600 Melting Pt: 176 Storage Temperature: Ambient |
MDL Number:
MFCD00149613 CAS Number: 10035-04-8 EINECS: 233-140-8 Merck Index: 13,01660 |
Specification Test Results
Appearance | White solid |
Fluoride | ≤ 0.004 % |
Lead | ≤ 5 mg/kg |
Grade | USP/FCC |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...