Copper(II) sulfate, anhydrous

Supplier: RPI
Warning

Synonyms: Cupric sulphate

C12150-100.0 C12150-500.0
76345-332EA 87.95 USD
76345-332 76345-334
Copper(II) sulfate, anhydrous
Copper(II) sulphate
Formula: CuSO₄
MW: 159.61 g/mol
Melting Pt: 200°C
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Grayish white powder
Density ≥ 3.6 g/ml
Assay ≥ 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR