Calcium-D gluconate, monohydrate
Supplier: RPI
Synonyms:
Calcium Gluconate, Calcium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconate Calcium, D-Gluconic acid calcium salt
C30050-1000.0
C30050-250.0
76345-370EA
85.85
USD
76345-370
76345-372
Calcium-D gluconate, monohydrate
Calcium-D gluconate
Additional calcium (Ca2+) source in plant tissue culture media.
Formula:
C12H22CaO14-H2O MW: 430.38 g/mol Melting Pt: 178°C Storage Temperature: Ambient |
MDL Number:
MFCD00064209 CAS Number: 299-28-5 EINECS: 206-075-8 |
Specification Test Results
Appearance | White crystalline powder |
Appearance of Solution | Conform |
Loss on Drying | ≤ 0.5 % |
Solubility | Sparingly soluble in water, freely soluble in boiling water |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...