Human Recombinant CD155 (from HEK293)

Supplier: ACROBIOSYSTEMS INC MS

CD5H82E3200UG CD5H82E325UG
103738-178EA 1685.38 USD
103738-178 103738-180
Human Recombinant CD155 (from HEK293)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/Peptide Name CD155
Environmentally Preferable
Protein Synonyms TAGE4|PVS|FLJ25946|CD155|NECL5|PVR|HVED
Protein/Peptide Type Recombinant
Species Human
Source HEK293 cells
Protease-free Y
Animal-Free Y
Nuclease-free Y
Carrier-Free Y
Endotoxin Level Low
Endotoxin Content Less than 1.0 EU per μg by the LAL method.
Biological Activity Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human TIGIT, Mouse IgG2a Fc tag, low endotoxin (Cat. No. TIT-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD155, His,Avitag (Cat. No. CD5-H82E3) with a linear range of 0.078-1.25 μg/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human CD155, His,Avitag (Cat. No. CD5-H82E3) at 5 μg/mL (100 μL/well) via streptavidin can bind Human TIGIT, Fc Tag (Cat. No. TIT-H5254) with a linear range of 0.1-2 ng/mL (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human CD155, His,Avitag (Cat. No. CD5-H82E3) at 5 μg/mL (100 μL/well) via streptavidin can bind Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) with a linear range of 0.1-1 ng/mL (Routinely tested).
Purity >95%
Formulation Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Molecular Weight 38.8 kDa
Storage Conditions –20 °C
Shipping Temperature Powder,RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR