2-tert-Butyldimethylsiloxybut-3-yne 97%, Sigma-Aldrich®

Warning

Synonyms: tert-Butyl-dimethyl-(methyl-prop-2-ynloxy)silane

M-Clarity™ quality level = MQ600 (more info.)

667579-1G 667579-10G
220087-304EA 71.4 USD
220087-304 220087-303
2-tert-Butyldimethylsiloxybut-3-yne 97%, Sigma-Aldrich®
2-tert-Butyldimethylsiloxybut-3-yne
Formula: C₁₀H₂₀OSi
MW: 184.35 g/mol
Flash Pt: 39 °C
MDL Number: MFCD08276883
CAS Number: 125494-93-1

Order Now

Specification Test Results

Appearance (Color) Colorless to Yellow
Appearance (Form) Liquid
Infrared spectrum Conforms to Structure
Purity (GC) ≥ 96.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR