Sodium pyruvate (synthetic) ≥99.5% (by HPLC), pharma grade, SAFC®

Warning

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

M-Clarity™ quality level = MQ500

80443-250G 80443-1KG
102292-262EA 82.62 USD
102292-262 102292-528
Sodium pyruvate (synthetic) ≥99.5% (by HPLC), pharma grade, SAFC®
Sodium pyruvate

Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.

Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: >300 °C (lit.)
MDL Number: MFCD00002586
CAS Number: 113-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR