Corning® 3D Clear Washing Buffer 10X, Corning
Supplier: Corning
5740 5741
76423-698CS 137.85 USD
76423-698 76423-700
Corning® 3D Clear Washing Buffer 10X, Corning
Buffers Cell Culture Buffers
Corning 3D Clear Washing Buffer 10X is used for the Corning 3D Clear immunolabeling process.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent lables
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
The Corning 3D Clear Washing Buffer 10X is used for the Corning 3D Clear immunolabeling process. This solution is a concentrate and is comprised of: PPBS with 0.2% Tween-20 and 10µg/mL heparin. This solution should be diluted to 1X concentration with PBS prior to use
Precaución: For best results, refrigerate after opening.