6-Benzylaminopurine 98,0

Supplier: RPI
Warning

Synonyms: 6-BAP, 6-Benzyladenine, N6-Benzyladenine, Benzyl(purin-6-yl)amine

B30070-25.0 B30070-5.0
76345-124EA 266.27 USD
76345-124 76345-126
6-Benzylaminopurine 98,0
6-Benzylaminopurine

Cytokine growth regulator.


Precaución: Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₂H₁₁N₅
MW: 225.25 g/mol
Melting Pt: 229…233 °C
MDL Number: MFCD00005572
CAS Number: 1214-39-7
EINECS: 214-927-5

Order Now

Specification Test Results

Appearance: White Crystalline Powder
Assay: ≥ 98.0 %
Loss on Drying: ≤ 1.0 %
Melting Point: 230 - 235 oC
Residue on Ignition: ≤ 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR