L-Arginine hydrochloride 98.0-101.5%
Supplier: RPI
Synonyms:
H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride
A50400-100.0
A50400-1000.0
A50400-3000.0
A50400-500.0
A50400-5000.0
76345-006EA
47.33
USD
76345-006
76345-008
76345-010
76345-012
76345-014
L-Arginine hydrochloride 98.0-101.5%
L-Arginine hydrochloride
Precaución: Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
C₆H₁₅ClN₄O₂ MW: 210.66 g/mol Melting Pt: 226 …230 °C |
MDL Number:
MFCD00064550 CAS Number: 1119-34-2 EINECS: 214-275-1 Merck Index: 13,00785 |
Specification Test Results
Appearance: | White crystalline powder |
Assay: | 98.5 - 101.5 % |
Heavy Metals: | ≤ 0.002 % |
Residue on Ignition: | ≤ 0.1 % |
Sulfate: | ≤ 0.03 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...