Amphotericin B solution 250 µg/ml
Supplier: RPI
Synonyms:
(1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
A40035-20.0
A40035-50.0
76344-920EA
39.4
USD
76344-920
76344-922
Amphotericin B solution 250 µg/ml
Amphotericin B
Plant cell culture tested.
- Polyene antifungal antibiotic produced by Streptomyces nodosus
- Interferes with the permeability of cell wall membrane of sensitive fungi and yeasts by binding to sterols
Precaución: Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
C₄₇H₇₃NO₁₇ MW: 924.08 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00877763 CAS Number: 1397-89-3 |
Specification Test Results
Appearance: | Clear yellow solution |
Concentration: | 250 µg/ml |
pH: | 6.5 - 8.5 |
Sterility: | Sterile Filtered |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...