Ampicillin trihydrate plant cell culture tested

Supplier: RPI
Danger

Synonyms: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

A40050-0.1 A40050-1.0 A40050-100.0 A40050-25.0 A40050-5.0 A40050-500.0
76344-938EA 26.86 USD
76344-938 76344-940 76344-942 76344-944 76344-946 76344-948
Ampicillin trihydrate plant cell culture tested
Ampicillin trihydrate

Plant cell culture tested.


  • Inhibitor of bacterial cell wall synthesis.
  • Highly active against gram negative bacteria.
  • Sensitive to β-Lactamase.

Formula: C₁₆H₁₉N₃O₄S·3H₂O
MW: 403.45 g/mol
Melting Pt: 198 …200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072036
CAS Number: 7177-48-2

Order Now

Specification Test Results

Appearance White Crystalline Powder
Potency 900 - 1050 µg/mg
Specific Optical Rotation +280 - +305 °
Water (KF) 12.0 - 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR