Amoxicillin trihydrate plant cell culture tested

Supplier: RPI
Danger

Synonyms: 6-[2-Amino-2-(4-hydroxyphenyl)-acetyl]amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxicillin Ttrihydrate, (2S,5R,6S)-6-((S)-2-amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

A40020-10.0 A40020-25.0
76344-904EA 90.15 USD
76344-904 76344-906
Amoxicillin trihydrate plant cell culture tested
Amoxicillin trihydrate

Plant cell culture tested.


  • Inhibitor of bacterial cell wall synthesis.
  • Highly active against gram negative bacteria. Highly active against gram negative bacteria.
  • Sensitive to β-Lactamase.

Formula: C₁₆H₁₉N₃O₅S
MW: 365.40 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00072029
CAS Number: 61336-70-7

Order Now

Specification Test Results

Appearance White or almost White crystalline powder
Solubility Colorless to light yellow
Assay (Anhydrous Substance) 900 - 1050 µg/mg
Dimethylaniline <0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR