Amikacin disulphate

Supplier: RPI

Synonyms: (S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)

A20540-100.0 A20540-25.0 A20540-50.0
76344-800EA 358.27 USD
76344-800 76344-802 77510-168
Amikacin disulphate
Amikacin disulphate

Semisynthetic, aminoglycoside antibiotic derived from Kanamycin A.


  • Range of activity is similar to Gentamicin.
  • Active against many strains of Gram negative bacteria.

Formula: C₂₂H₄₃N₅O₁₃·2H₂SO₄
MW: 781.77 g/mol
Melting Pt: 234 -236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00167475
CAS Number: 39831-55-5
EINECS: 254-648-6

Order Now

Specification Test Results

Appearance White Crystalline Powder
Assay (Dried basis) 674 - 786 µg/mg
Loss on Drying ≤ 13.0 %
pH 2.0 - 4.0
Residue on Ignition ≤ 1.0 %
Specific Rotation +76 - +84 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR