4-Aminobenzoic acid ≥98.5%
Supplier: RPI
|
Warning
|
Synonyms:
Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1
A20050-100.0
A20050-25.0
76344-650EA
220.73
USD
76344-650
76344-652
4-Aminobenzoic acid ≥98.5%
4-Aminobenzoic acid
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Melting Pt: 180…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007894 CAS Number: 150-13-0 EINECS: 205-753-0 |
Specification Test Results
Appearance | White crystalline powder |
Assay | ≥ 98.5 % |
Loss on Drying | ≤ 0.20 % |
Melting Point | 184 - 191 oC |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...