Adenine sulphate ≥98.0%

Supplier: RPI

Synonyms: 6-Aminopurine hemisulphate salt, 7H-Purin-6-amine hemisulphate, Adenine hemisulphate salt

A12575-100.0 A12575-25.0 A12575-500.0
76344-590EA 289.19 USD
76344-590 76344-592 76344-594
Adenine sulphate ≥98.0%
Adenine sulphate

Adenine Sulfate [Adenine Hemisulfate Salt]


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₅H₅N₅·½H₂SO₄
MW: 202.2 g/mol
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00213655
CAS Number: 321-30-2
EINECS: 206-286-5

Order Now

Specification Test Results

Appearance White to Off White Crystalline Powder
Purity (HPLC) ≥ 98.0 %
Loss on Drying ≤10.0 %
Residue on Ignition ≤ 0.1 %
Heavy Metals ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR