Adenosine 5'-monophosphate monohydrate
Supplier: RPI
Synonyms:
A-5′-P, AMP, Adenosine-5'-monophosphoric acid monohydrate, 5′-AMP, 5′-Adenylic acid
A11240-1.0
A11240-10.0
A11240-5.0
76344-512EA
25.11
USD
76344-512
76344-514
76344-516
Adenosine 5'-monophosphate monohydrate
Adenosine 5'-monophosphate monohydrate
AMP [Adenosine-5-Monophosphate, Free Acid]
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
C₁₀H₁₄N₅O₇P·1H₂O MW: 365.24 g/mol |
MDL Number:
MFCD00149360 CAS Number: 18422-05-4 EINECS: 200-500-0 |
Specification Test Results
Appearance | Solid |
Molar Absorptivity (@260nm - pH 7.5) | 14.9 - 15.9 X103L/mole/cm |
Ratio @ pH 7.5 A250/A260 | 0.75 - 0.81 |
Water Content | ≤ 8.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...