Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Supplier: RPI
Synonyms:
Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
A11220-1.0
A11220-10.0
A11220-5.0
76344-506EA
36.47
USD
76344-506
76344-508
76344-510
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
ADP [Adenosine-5'-diphosphate, disodium salt dihydrate]
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
C₁₀H₁₃N₅Na₂O₁₀P₂ MW: 471.167 g/mol |
MDL Number:
MFCD00066635 CAS Number: 16178-48-6 EINECS: 240-314-7 |
Specification Test Results
Appearance | White Powder |
Molar Absorptivity (@260nm - pH 7.5) | 14.9 - 15.9 X103L/mole/cm |
Ratio @ pH 7.5 A250/A260 | 0.75 - 0.81 |
Sodium Contents | 8.0 - 12.0 |
Water Content | ≤ 8 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...