Capillary Cassettes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G7100-60002 G1600-60002
101407-660EA 724.66 USD
101407-660 101443-154 101443-160 101443-162 101443-164 101443-166 101443-168 101443-170 101484-546 101484-556 101484-558 101484-562 101484-566 101484-570 103577-718 AGG7100-60002 AGG1600-60002
Capillary Cassettes, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Quick-change capillary cassettes that enable capillary exchange in less than a minute.
  • The quick-change cassette allows capillary exchange in less than a minute
  • The cassette and interfaces accept all commercially available capillaries with a 365 µm OD
Agilent provides quick-change capillary cassettes that enable capillary exchange in less than a minute. The color code of the alignment interface must match with the color code of the capillary's built-in alignment stopper.