IGEPAL® CA-630, Ultrapure

Danger

Synonyms: Octylphenyl-polyethylene glycol

J19628-K2 J19628-AP
103728-104EA 150.77 USD
103728-104 103728-144
IGEPAL® CA-630, Ultrapure
IGEPAL® CA-630
Formula: (C₂H₄O)nC₁₄H₂₂O
Boiling Pt: >250 °C
Melting Pt: ~5 °C
Density: 1.061
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00132505
CAS Number: 9002-93-1
Merck Index: 14,06761

Order Now

Specification Test Results

Form Clear, slightly yellow-green viscous liquid
Optical Rotation 1.06 ± 0.005

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR