Sodium cholate 97+% (dry basis), Ultrapure

Warning

Synonyms: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, Sodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoate, Cholic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, sodium salt

J13630-09 J13630-22 J13630-36 J13630-A1
103728-068EA 66.88 USD
103728-068 103728-070 103728-072 103728-074
Sodium cholate 97+% (dry basis), Ultrapure
Sodium cholate
Formula: C₂₄H₃₉NaO₅
MW: 430.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150749
CAS Number: 361-09-1
EINECS: 206-643-5

Order Now

Specification Test Results

Form White powder
Assay (unspecified) ≥ 97.0% (dry basis)
Identification (FTIR) Conforms
Loss on Drying ≤ 5.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR