LipidSpot™ 488 lipid droplet stains
Supplier: Biotium
70065 70065-T
76221-448EA 551.87 USD
76221-448 76221-660
LipidSpot™ 488 lipid droplet stains
LipidSpot™
LipidSpot™ stains are fluorescent dyes that rapidly stain lipid droplets in live or fixed cells, with no wash step and minimal background.
  • Rapidly and specifically stain lipid droplets
  • Minimal background, no wash required
  • Stain live or fixed cells, or fix and permeabilize after staining
  • Available with green or red/far-red fluorescence
  • LipidSpot™ 488 validated in super-resolution imaging by SIM
  • Supplied at 1000X in DMSO
Intracellular lipid droplets are cytoplasmic organelles involved in the storage and regulation of triglycerides and cholesterol esters. LipidSpot™ dyes are fluorogenic neutral lipid stains that rapidly accumulate in lipid droplets, where they become brightly fluorescent. The dyes can be used to stain lipid droplets in both live and fixed cells, with no wash step required. Cells also can be fixed and permeabilized after staining. LipidSpot™ stains show minimal background staining of cellular membranes or other organelles, unlike traditional dyes like Nile Red
LipidSpot™ 488 has excitation around 430 nm, and can be excited equally well at 405 nm or 488 nm. In cells, it stains lipid droplets with bright green fluorescence detectable in the FITC channel. LipidSpot™ 488 has been validated in super-resolution imaging by SIM.
In yeast, LipidSpot™ 488 stains intracellular membranes, but LipidSpot™ 610 does not. In bacteria, both LipidSpot™ dyes can stain gram-positive but not gram-negative strains. See our Cellular Stains Table for more information on how our dyes stain various organisms.
Order Now
Specification Test Results
Abs/Em 427/585 nm (in vegetable oil or cells)
Detection channel FITC, GFP