PIG® Lab and Chemical Cleanup Kits, New Pig
Supplier: New Pig
KIT5007 KIT5009
76326-534EA 38.58 USD
76326-534 76326-536
PIG® Lab and Chemical Cleanup Kits, New Pig
Spill Control Materials
Be prepared for small spills in the laboratory with our grab-and-go clean up kit for acids, bases and chemicals.
  • Compact design lets you store a spill kit virtually anywhere you need one for fast response
  • Easy-open, resealable bag protects kit contents from dirt and moisture
Single-use kit is faster and more efficient than dragging a large spill kit to a small cleanup; safer and more effective than using rags or paper towels.
Only PIG Spill Kits feature PIG Absorbents.
Información de suministro: Includes hazmat absorbent pads, acid neutralizing loose absorbent and base neutralizing loose absorbent to clean up most acids, bases and chemicals (absorbs up to 25 oz.). Disposable gloves, mini dustpan and disposal bag also included to aid cleanup.