NanoXact Silver-shelled Gold Nanospheres 1 mg/ml, nanoComposix
Supplier: NANOCOMPOSIX, INC.
BMCH20-10M BMCH20-1M BMCH20-30M BMCH20-5M
103820-100EA 1865 USD
103820-100 103820-102 103820-104 103820-106 103820-108 103820-110 103820-112 103820-114 77680-220 77680-222 77680-224 77680-226
NanoXact Silver-shelled Gold Nanospheres 1 mg/ml, nanoComposix
Beads Microcarriers
Bimetallic Silver-Shelled Gold Nanospheres with displaceable citrate surface.
  • Bimetallic Nanospheres with unqiue plasmonic properties.
NanoXact, Silver to shelled Gold Nanospheres with a citrate surface. Bimetallic core to shell nanoparticles are of interest for plasmonic studies and as highly to characterized constructs for single particle elemental analysis.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, and UV-Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Información de suministro: Ships at ambient temperatures
Precaución: Do not freeze. Store at 2 to 8 °C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.